Assay Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026231.2 | NP_080507.2 | 6 - 7 | 1542 | 82 | |
| GenBank mRNA | AK013967.1 | - | 3 - 4 | 394 | 82 | |
| AK017059.1 | - | 4 - 5 | 817 | 82 | ||
| AK020021.1 | - | 5 - 6 | 1152 | 82 | ||
| AK029724.1 | - | 6 - 7 | 1542 | 83 | ||
| AK033409.1 | - | 6 - 7 | 1580 | 82 | ||
| AK044441.1 | - | 6 - 7 | 1550 | 82 | ||
| BC047136.1 | - | 6 - 7 | 1542 | 6311866 | 82 | |
| BC061240.1 | - | 6 - 7 | 1538 | 6743615 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gstcd
Genomic Map