Assay Details
| Gene Symbol: Gstcd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026231.2 | NP_080507.2 | 8 - 9 | 1714 | 126 | |
| GenBank mRNA | AK013967.1 | - | 5 - 6 | 566 | 126 | |
| AK017059.1 | - | 6 - 7 | 989 | 126 | ||
| AK020021.1 | - | 7 - 8 | 1324 | 126 | ||
| AK029724.1 | - | 8 - 9 | 1715 | 127 | ||
| AK033409.1 | - | 8 - 9 | 1752 | 126 | ||
| BC047136.1 | - | 8 - 9 | 1714 | 6311866 | 126 | |
| BC061240.1 | - | 8 - 9 | 1710 | 6743615 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gstcd
Genomic Map