Assay Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK041195.1 | - | 1 - 2 | 153 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK041195.1 transcripts.
Set Membership: |
|

Slc35a3
Genomic Map