Assay Details
| Gene Symbol: Lrrc28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175124.5 | NP_780333.3 | 5 - 6 | 509 | 79 | |
| GenBank mRNA | AK077270.1 | - | 5 - 6 | 505 | 79 | |
| AK159417.1 | - | 5 - 6 | 478 | 79 | ||
| BC086694.1 | - | 5 - 6 | 400 | 30697866 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc28
Genomic Map