Assay Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290514.1 | NP_001277443.1 | 1 - 2 | 134 | 115 | |
| NM_027409.5 | NP_081685.1 | 1 - 2 | 134 | 115 | ||
| NR_110964.1 | 1 - 2 | 134 | 115 | |||
| GenBank mRNA | AK088275.1 | - | 1 - 2 | 71 | 115 | |
| AK139795.1 | - | 1 - 2 | 78 | 115 | ||
| BC018329.1 | - | 1 - 2 | 35 | 3962906 | 115 | |
| BC085100.1 | - | 1 - 2 | 42 | 6391017 | 115 | |
| CJ046213.1 | - | 1 - 2 | 135 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139795.1, BC018329.1, BC085100.1, NM_001290514.1, NM_027409.5 transcripts.
Set Membership: |
|

Mospd1
Genomic Map