| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400018908.mm.1 |
NM_001160326.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018908.mm.1 |
NM_001160326.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018908.mm.1 |
TC1400002447.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018907.mm.1 |
NM_001160326.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018907.mm.1 |
NM_001160326.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018907.mm.1 |
NR_027699.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018907.mm.1 |
NR_027699.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018907.mm.1 |
TC1400002447.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400018906.mm.1 |
NM_001160326.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018906.mm.1 |
NM_001160326.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018906.mm.1 |
NR_027699.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018906.mm.1 |
NR_027699.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400018906.mm.1 |
TC1400002447.mm.1 |
MTA TC |