Assay Details
Gene Symbol: Nek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010892.3 | NP_035022.2 | 4 - 5 | 807 | 82 | |
GenBank mRNA | AF007247.1 | - | 4 - 5 | 721 | 82 | |
AF013166.1 | - | 4 - 5 | 691 | 82 | ||
AK077627.1 | - | 4 - 5 | 771 | 82 | ||
AK147072.1 | - | 4 - 5 | 783 | 82 | ||
AK164467.1 | - | 4 - 5 | 750 | 82 | ||
AK169330.1 | - | 5 - 6 | 698 | 82 | ||
AK170179.1 | - | 4 - 5 | 727 | 82 | ||
AK171277.1 | - | 4 - 5 | 727 | 82 | ||
AK171387.1 | - | 4 - 5 | 727 | 82 | ||
BC010302.1 | - | 4 - 5 | 729 | 3597227 | 82 | |
BC011316.1 | - | 4 - 5 | 727 | 3674001 | 82 | |
BC057576.1 | - | 4 - 5 | 747 | 6810914 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
