Assay Details
| Gene Symbol: Stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198659.2 | NP_941061.1 | 10 - 11 | 1263 | 153 | |
| GenBank mRNA | AK046955.1 | - | 10 - 11 | 1264 | 153 | |
| AK161389.1 | - | 10 - 11 | 1258 | 153 | ||
| BC138108.1 | - | 10 - 11 | 1302 | 8861127 | 153 | |
| BC138109.1 | - | 10 - 11 | 1302 | 8861128 | 153 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stpg2
Genomic Map