Assay Details
| Gene Symbol: Steap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103156.2 | NP_001096626.1 | 2 - 3 | 262 | 105 | |
| NM_001103157.2 | NP_001096627.1 | 2 - 3 | 617 | 105 | ||
| NM_001285469.1 | NP_001272398.1 | 2 - 3 | 171 | 105 | ||
| NM_028734.5 | NP_083010.2 | 2 - 3 | 319 | 105 | ||
| GenBank mRNA | AK052981.1 | - | 2 - 3 | 321 | 105 | |
| AK086787.1 | - | 2 - 3 | 289 | 105 | ||
| AK145468.1 | - | 2 - 3 | 208 | 105 | ||
| AK166162.1 | - | 2 - 3 | 285 | 105 | ||
| AK166607.1 | - | 2 - 3 | 208 | 105 | ||
| BC043459.1 | - | 2 - 3 | 171 | 105 | ||
| BC150881.1 | - | 2 - 3 | 178 | 9087792 | 105 | |
| BC150889.1 | - | 2 - 3 | 178 | 9087800 | 105 | |
| BC150997.1 | - | 229 | 9087910 | 105 | ||
| BC151008.1 | - | 229 | 9087921 | 105 | ||
| CJ122203.1 | - | 2 - 3 | 266 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK052981.1, BC150881.1, BC150889.1, BC150997.1, BC151008.1, NM_001103156.2, NM_001103157.2, NM_001285469.1, NM_028734.5 transcripts.
Set Membership: |
|

Steap2
Genomic Map