Assay Details
| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031375.4 | NP_113552.3 | 1 - 2 | 207 | 104 | |
| GenBank mRNA | AB047544.1 | - | 1 - 2 | 163 | ||
| AK144756.1 | - | 1 - 2 | 203 | 104 | ||
| BC004034.1 | - | 194 | 3495639 | 104 | ||
| CJ154012.1 | - | 1 - 2 | 206 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB047544.1, AK144756.1, BC004034.1 transcripts.
Set Membership: |
|

Ngrn
Genomic Map