Assay Details
| Gene Symbol: Tmem126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026734.1 | NP_081010.1 | 3 - 4 | 421 | 86 | |
| GenBank mRNA | AK003201.1 | - | 3 - 4 | 422 | 86 | |
| BC049680.1 | - | 3 - 4 | 421 | 6596401 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313827 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313825 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313824 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676063 | AK003201.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676063 | BC049680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676063 | NM_026734.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676063 | AK003201.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676063 | BC049680.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P676063 | NM_026734.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605720 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390397 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 412576 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254819 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121008 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676063 | AK003201.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676063 | BC049680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676063 | NM_026734.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676063 | AK003201.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676063 | BC049680.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P676063 | NM_026734.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313821 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313827 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313838 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313825 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313824 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313834 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313807 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313823 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313828 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313817 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313815 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313819 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313839 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313855 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313812 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313806 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313866 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313813 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313809 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313852 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313851 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313829 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313816 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313810 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313857 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313805 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25313847 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06277003 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121008 | - | - |
| Whole Mouse Genome G4122A | A_52_P676063 | AK003201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676063 | BC049680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676063 | NM_026734.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676063 | AK003201.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676063 | BC049680.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P676063 | NM_026734.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121008 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06233159 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06233171 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06233151 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06233129 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313828 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313824 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313827 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25313825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313785 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25313825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313863 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06233127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313849 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313796 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06233120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313884 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313842 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06646630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313888 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313851 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06425664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05956736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313866 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313869 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313825 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313861 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06145050 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313865 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06278962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06233171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313867 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313806 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313827 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06233129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313819 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313852 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313833 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313836 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313821 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313885 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313847 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313829 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06339180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313854 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06626150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313878 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313792 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313850 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06233168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06278960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313872 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313844 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313855 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25313779 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10565522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10565521 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453739_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10565519 | - | - |
| Mouse Genome 430 2.0 Array | 1453739_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453739_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700031448.mm.1 | NM_026734.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700031448.mm.1 | NM_026734.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700031448.mm.1 | TC0700003727.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700031449.mm.1 | NM_026734.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700031449.mm.1 | NM_026734.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700031449.mm.1 | TC0700003727.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453739_at | - | - |
| Mouse Expression Set 430 | 1453739_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453739_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453739_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 301828 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259294 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1259294 | - | - |

Tmem126b
Genomic Map