Assay Details
| Gene Symbol: Sorbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271407.1 | NP_001258336.1 | 18 - 19 | 2093 | 72 | |
| NM_001271408.1 | NP_001258337.1 | 18 - 19 | 1977 | 72 | ||
| NM_001271409.1 | NP_001258338.1 | 7 - 8 | 958 | 72 | ||
| NM_011366.3 | NP_035496.1 | 18 - 19 | 1980 | 72 | ||
| GenBank mRNA | AK029703.1 | - | 20 - 21 | 2092 | 72 | |
| AK165689.1 | - | 18 - 19 | 1987 | 72 | ||
| BC030933.1 | - | 17 - 18 | 1794 | 4920799 | 72 | |
| U58889.1 | - | 1 - 2 | 262 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sorbs3
Genomic Map