Assay Details
| Gene Symbol: Mospd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290514.1 | NP_001277443.1 | 4 - 5 | 683 | 126 | |
| NR_110964.1 | 4 - 5 | 683 | 126 | |||
| GenBank mRNA | AK088275.1 | - | 4 - 5 | 620 | 126 | |
| AK139795.1 | - | 4 - 5 | 627 | 126 | ||
| BC018329.1 | - | 4 - 5 | 584 | 3962906 | 126 | |
| BC085100.1 | - | 4 - 5 | 591 | 6391017 | 126 | |
| BC100442.1 | - | 4 - 5 | 571 | 30501889 | 126 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mospd1
Genomic Map