Assay Details
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175127.2 | NP_780336.1 | 3 - 4 | 535 | 95 | |
| GenBank mRNA | AK076211.1 | - | 3 - 4 | 541 | 95 | |
| AK077653.1 | - | 3 - 4 | 548 | 95 | ||
| AK122295.1 | - | 3 - 4 | 520 | 95 | ||
| AK162091.1 | - | 3 - 4 | 504 | 95 | ||
| AK162188.1 | - | 3 - 4 | 416 | 95 | ||
| BC055458.1 | - | 2 - 3 | 237 | 95 | ||
| BC094542.1 | - | 3 - 4 | 535 | 6408744 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fbxo28
Genomic Map