Assay Details
Gene Symbol: Casp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK011710.1 | - | 2 - 3 | 94 | 96 | |
AK146363.1 | - | 2 - 3 | 104 | 96 | ||
AK164321.1 | - | 2 - 3 | 70 | 96 | ||
AK165998.1 | - | 2 - 3 | 81 | 96 | ||
AK168842.1 | - | 2 - 3 | 102 | 96 | ||
BC002022.1 | - | 2 - 3 | 90 | 3491126 | 96 | |
BY070840.1 | - | 2 - 3 | 130 | 96 | ||
Y13087.1 | - | 1 - 2 | 58 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK011710.1, AK146363.1, AK164321.1, AK165998.1, AK168842.1, BC002022.1, Y13087.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
