Assay Details
| Gene Symbol: Cyp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020010.2 | NP_064394.2 | 2 - 3 | 663 | 99 | |
| GenBank mRNA | AK028355.1 | - | 2 - 3 | 356 | 99 | |
| AK028815.1 | - | 2 - 3 | 420 | 99 | ||
| AK031059.1 | - | 2 - 3 | 396 | 99 | ||
| AK075764.1 | - | 2 - 3 | 396 | 99 | ||
| AK076983.1 | - | 2 - 3 | 396 | 99 | ||
| AK202785.1 | - | 1 - 2 | 109 | 99 | ||
| AK208162.1 | - | 1 - 2 | 88 | 100 | ||
| BC031813.1 | - | 2 - 3 | 390 | 4193738 | 99 | |
| CB524876.1 | - | 2 - 3 | 663 | 99 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp51
Genomic Map