Assay Details
| Gene Symbol: Cyp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020010.2 | NP_064394.2 | 3 - 4 | 832 | 88 | |
| GenBank mRNA | AF166266.1 | - | 3 - 4 | 584 | 88 | |
| AK028355.1 | - | 3 - 4 | 525 | 88 | ||
| AK028815.1 | - | 3 - 4 | 589 | 88 | ||
| AK031059.1 | - | 3 - 4 | 565 | 88 | ||
| AK075764.1 | - | 3 - 4 | 565 | 88 | ||
| AK076983.1 | - | 3 - 4 | 565 | 88 | ||
| AK208162.1 | - | 2 - 3 | 260 | 88 | ||
| BC031813.1 | - | 3 - 4 | 559 | 4193738 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyp51
Genomic Map