| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700009690.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009690.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009690.mm.1 |
TC0700001101.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009697.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009697.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009697.mm.1 |
TC0700001101.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009695.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009695.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009695.mm.1 |
TC0700001101.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009696.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009696.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009696.mm.1 |
TC0700001101.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009700.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009700.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009700.mm.1 |
TC0700001101.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009693.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009693.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009693.mm.1 |
TC0700001101.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009694.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009694.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009694.mm.1 |
TC0700001101.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700009698.mm.1 |
NM_001199224.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009698.mm.1 |
NM_001199224.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700009698.mm.1 |
TC0700001101.mm.1 |
MTA TC |