Assay Details
| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197997.2 | NP_932114.2 | 3 - 4 | 333 | 80 | |
| GenBank mRNA | BC079891.1 | - | 3 - 4 | 333 | 80 | |
| BC082599.1 | - | 3 - 4 | 217 | 80 | ||
| BC085187.1 | - | 3 - 4 | 189 | 80 | ||
More Information
Set Membership: |
|

Zgrf1
Genomic Map