| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300010449.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010449.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010449.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010452.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010452.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010452.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010453.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010453.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010453.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010448.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010448.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010448.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010451.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010451.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010451.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010450.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010450.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010450.mm.1 |
TC0300001289.mm.1 |
MTA TC |