Assay Details
| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197997.2 | NP_932114.2 | 10 - 11 | 3292 | 73 | |
| GenBank mRNA | AK046429.1 | - | 5 - 6 | 1018 | 73 | |
| BC079891.1 | - | 9 - 10 | 3168 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK046429.1 transcripts.
Set Membership: |
|

Zgrf1
Genomic Map