| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300010461.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010461.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010461.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010470.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010470.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010470.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010474.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010474.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010474.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010467.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010467.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010467.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010468.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010468.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010468.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010462.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010462.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010462.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010463.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010463.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010463.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010469.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010469.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010469.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010472.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010472.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010472.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010465.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010465.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010465.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010466.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010466.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010466.mm.1 |
TC0300001289.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300010473.mm.1 |
NM_197997.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010473.mm.1 |
NM_197997.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300010473.mm.1 |
TC0300001289.mm.1 |
MTA TC |