| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2180181 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
104692 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
44039 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
399887 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
63230 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
823336 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457047 |
AK049095.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457047 |
AK076310.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457047 |
AK088830.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
457047 |
NM_139294.5 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
98884 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
177675 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2180181 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06243593 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P108369 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25236814 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06148054 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25236826 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25236818 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25236816 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25236824 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06148051 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06148047 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25236823 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25236812 |
- |
- |