| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1437424_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10496748 |
- |
- |
| Mouse Genome 430 2.0 Array |
1437424_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1437424_PM_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
214804_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
214804_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0300012005.mm.1 |
NM_001166064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012005.mm.1 |
NM_001166064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012005.mm.1 |
TC0300001486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300012006.mm.1 |
NM_001166064.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012006.mm.1 |
NM_001166064.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300012006.mm.1 |
TC0300001486.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1437424_PM_at |
- |
- |
| Mouse Expression Set 430 |
1437424_at |
- |
- |
| Murine Genome U74v2 Set |
167776_i_at |
- |
- |
| Murine Genome U74v2 Set |
129316_at |
- |
- |
| Murine Genome U74v2 Set |
167776_PM_i_at |
- |
- |
| Murine Genome U74v2 Set |
129316_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1437424_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1437424_at |
- |
- |