Assay Details
| Gene Symbol: Nynrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040072.1 | NP_001035161.1 | 1 - 2 | 312 | 57 | |
| GenBank mRNA | BC151043.1 | - | 2 - 3 | 334 | 9087956 | 57 |
More Information
Updated Mapping Information
Set Membership: |
|

Nynrin
Genomic Map