Assay Details
| Gene Symbol: Nynrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040072.1 | NP_001035161.1 | 5 - 6 | 2599 | 71 | |
| GenBank mRNA | AK220380.1 | - | 3 - 4 | 448 | 71 | |
| BC151043.1 | - | 6 - 7 | 2621 | 9087956 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nynrin
Genomic Map