Assay Details
| Gene Symbol: Nynrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040072.1 | NP_001035161.1 | 6 - 7 | 2702 | 81 | |
| GenBank mRNA | AK220380.1 | - | 4 - 5 | 551 | 81 | |
| BC151043.1 | - | 7 - 8 | 2724 | 9087956 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nynrin
Genomic Map