Assay Details
| Gene Symbol: Nynrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040072.1 | NP_001035161.1 | 7 - 8 | 2799 | 67 | |
| GenBank mRNA | AK220380.1 | - | 5 - 6 | 648 | 67 | |
| BC151043.1 | - | 8 - 9 | 2821 | 9087956 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042984 | BC151043.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2042984 | NM_001040072.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627628 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 742754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 50933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658351 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 626883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123690 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 825886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601605 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 481527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 795662 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132129 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145198 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042984 | BC151043.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2042984 | NM_001040072.1 | mouse_transcriptome |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11201114 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11715160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11395888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11477745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620271 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11201106 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620317 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11201088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620332 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620288 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11715159 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11201105 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11138035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11807906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11765237 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620266 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11715147 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29620331 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10415409 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415410 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415412 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10415414 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439064_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415413 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415411 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10415408 | - | - |
| Mouse Genome 430 2.0 Array | 1439064_at | - | - |
| Mouse Genome 430 2.0 Array | 1439064_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277154_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277154_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400006474.mm.1 | NM_001040072.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006474.mm.1 | NM_001040072.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006474.mm.1 | TC1400000796.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400006475.mm.1 | NM_001040072.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006475.mm.1 | NM_001040072.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400006475.mm.1 | TC1400000796.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439064_at | - | - |
| Mouse Expression Set 430 | 1439064_PM_at | - | - |
| Murine Genome U74v2 Set | 103485_at | - | - |
| Murine Genome U74v2 Set | 103485_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439064_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439064_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439064_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439064_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 849785 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234939 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242910 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1234939 | - | - |

Nynrin
Genomic Map