Assay Details
| Gene Symbol: Nmi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001141948.1 | NP_001135420.1 | 7 - 8 | 1050 | 88 | |
| NM_001141949.1 | NP_001135421.1 | 8 - 9 | 1090 | 88 | ||
| NM_019401.2 | NP_062274.1 | 7 - 8 | 985 | 88 | ||
| GenBank mRNA | AF019249.1 | - | 7 - 8 | 893 | 88 | |
| AK146577.1 | - | 8 - 9 | 887 | 88 | ||
| BC002019.1 | - | 7 - 8 | 845 | 3590545 | 88 | |
| CT010361.1 | - | 6 - 7 | 744 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of type I interferon production
negative regulation of interferon-alpha biosynthetic process
negative regulation of interferon-beta biosynthetic process
negative regulation of innate immune response
positive regulation of protein K48-linked ubiquitination
protein binding
negative regulation of type I interferon production
negative regulation of interferon-alpha biosynthetic process
negative regulation of interferon-beta biosynthetic process
negative regulation of innate immune response
positive regulation of protein K48-linked ubiquitination
protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21216635 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21216637 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034705 | AK146577.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034705 | BC002019.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034705 | NM_001141948.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034705 | NM_001141949.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2034705 | NM_019401.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316095 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 446329 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390326 | BC058224.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390326 | BC080859.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 870165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91470 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814301 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 297516 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 391699 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474592 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 279177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289310 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 105640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125207 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128629 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122931 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174018 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034705 | AK146577.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034705 | BC002019.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034705 | NM_001141948.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034705 | NM_001141949.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2034705 | NM_019401.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216659 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216628 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216652 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216632 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21216625 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122931 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00368563 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174018 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04518031 | - | - |
| Whole Mouse Genome G4122A | A_51_P125067 | AK146577.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125067 | BC002019.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125067 | NM_001141948.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125067 | NM_001141949.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P125067 | NM_019401.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113579 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122931 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174018 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01952088 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01259707 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01259708 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21216635 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21216637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216529 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21216551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01649653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216673 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216620 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216671 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216669 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216666 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216551 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01956042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216652 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01339804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216632 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01916714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02181481 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216635 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01916719 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216609 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216655 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216604 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216549 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02113009 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01952088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01615866 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01971176 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216659 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216601 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21216680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02281362 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02058213 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10482527 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482518 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10482526 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425719_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10482517 | - | - |
| Mouse Genome 430 2.0 Array | 1425719_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1425719_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64685_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 64685_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200029975.mm.1 | NM_001141948.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029975.mm.1 | NM_001141948.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029975.mm.1 | NM_001141949.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029975.mm.1 | NM_001141949.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029975.mm.1 | NM_019401.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029975.mm.1 | NM_019401.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200029975.mm.1 | TC0200003528.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425719_a_at | - | - |
| Mouse Expression Set 430 | 1425719_PM_a_at | - | - |
| Murine Genome U74v2 Set | 101424_at | - | - |
| Murine Genome U74v2 Set | 101424_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425719_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425719_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425719_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425719_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 914220 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 520270 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000520270 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2755958 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2755958 | - | - |
| mouse-6 v1.1 (Illumina) | scl19291.9.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 520270 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl19291.9.1_25-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 520270 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2755958 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2755958 | - | - |

Nmi
Genomic Map