| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200015546.mm.1 |
NM_001289747.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015546.mm.1 |
NM_001289748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015546.mm.1 |
NM_177726.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015546.mm.1 |
TC0200001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015543.mm.1 |
NM_001289747.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015543.mm.1 |
NM_001289749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015543.mm.1 |
NM_177726.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015543.mm.1 |
TC0200001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015544.mm.1 |
NM_001289747.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015544.mm.1 |
TC0200001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015545.mm.1 |
NM_001289747.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015545.mm.1 |
NM_001289748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015545.mm.1 |
NM_001289749.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015545.mm.1 |
NM_177726.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015545.mm.1 |
TC0200001969.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015540.mm.1 |
NM_001289748.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015540.mm.1 |
TC0200001969.mm.1 |
MTA TC |