Assay Details
| Gene Symbol: Pfkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291071.1 | NP_001278000.1 | 7 - 8 | 863 | 101 | |
| NM_019703.4 | NP_062677.1 | 7 - 8 | 863 | 101 | ||
| GenBank mRNA | AF123533.1 | - | 7 - 8 | 828 | 101 | |
| AK004790.1 | - | 7 - 8 | 818 | 101 | ||
| AK049841.1 | - | 7 - 8 | 817 | 101 | ||
| AK076803.1 | - | 7 - 8 | 818 | 101 | ||
| AK078254.1 | - | 7 - 8 | 818 | 101 | ||
| AK098162.1 | - | 7 - 8 | 817 | 101 | ||
| AK152670.1 | - | 7 - 8 | 818 | 101 | ||
| AK154125.1 | - | 7 - 8 | 101 | |||
| AK165422.1 | - | 7 - 8 | 817 | 101 | ||
| AK165425.1 | - | 7 - 8 | 817 | 101 | ||
| AK170624.1 | - | 7 - 8 | 818 | 101 | ||
| AK171062.1 | - | 7 - 8 | 101 | |||
| AY779275.1 | - | 7 - 8 | 101 | |||
| AY779276.1 | - | 7 - 8 | 780 | 101 | ||
| BC006926.1 | - | 7 - 8 | 802 | 2655383 | 101 | |
| CT010268.1 | - | 7 - 8 | 771 | 101 | ||
| Y19008.1 | - | 7 - 8 | 820 | 101 | ||
More Information
Set Membership: |
|

Pfkp
Genomic Map