Assay Details
| Gene Symbol: Dph6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025675.4 | NP_079951.1 | 5 - 6 | 580 | 94 | |
| GenBank mRNA | AK006965.1 | - | 5 - 6 | 538 | 94 | |
| AK007021.1 | - | 5 - 6 | 583 | 94 | ||
| AK017595.1 | - | 5 - 6 | 536 | 94 | ||
| AK036021.1 | - | 5 - 6 | 616 | 94 | ||
| AK082013.1 | - | 5 - 6 | 617 | 94 | ||
| AK088160.1 | - | 5 - 6 | 582 | 94 | ||
| AK145789.1 | - | 5 - 6 | 540 | 94 | ||
| BC022995.1 | - | 5 - 6 | 605 | 5363234 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dph6
Genomic Map