| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0600017772.mm.1 |
NM_016806.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017772.mm.1 |
NM_182650.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017772.mm.1 |
NR_104468.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017772.mm.1 |
TC0600002265.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017769.mm.1 |
NM_016806.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017769.mm.1 |
NM_182650.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017769.mm.1 |
NR_104468.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017769.mm.1 |
TC0600002265.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017773.mm.1 |
NM_016806.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017773.mm.1 |
NM_182650.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017773.mm.1 |
NR_104468.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017773.mm.1 |
TC0600002265.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017771.mm.1 |
NM_016806.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017771.mm.1 |
NM_182650.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017771.mm.1 |
NR_104468.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017771.mm.1 |
TC0600002265.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0600017768.mm.1 |
NM_016806.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017768.mm.1 |
NM_182650.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017768.mm.1 |
NR_104468.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0600017768.mm.1 |
TC0600002265.mm.1 |
MTA TC |