Assay Details
| Gene Symbol: Dfna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016561.1 | - | 404 | 80 | ||
| BC051962.1 | - | 1 - 2 | 96 | 80 | ||
| BC086618.1 | - | 1 - 2 | 96 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016561.1 transcripts.
Set Membership: |
|

Dfna5
Genomic Map