Assay Details
| Gene Symbol: Dfna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018769.3 | NP_061239.1 | 3 - 4 | 466 | 65 | |
| GenBank mRNA | AF073309.1 | - | 3 - 4 | 467 | 65 | |
| AK016561.1 | - | 834 | 65 | |||
| AK089459.1 | - | 453 | 65 | |||
| AK155135.1 | - | 445 | 65 | |||
| AK171278.1 | - | 3 - 4 | 465 | 65 | ||
| BC051962.1 | - | 3 - 4 | 526 | 65 | ||
| BC086618.1 | - | 3 - 4 | 526 | 65 | ||
| BC132303.1 | - | 2 - 3 | 437 | 40130580 | 65 | |
More Information
Set Membership: |
|

Dfna5
Genomic Map