Assay Details
| Gene Symbol: Dfna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018769.3 | NP_061239.1 | 6 - 7 | 924 | 76 | |
| GenBank mRNA | AF073309.1 | - | 6 - 7 | 925 | 76 | |
| AK016561.1 | - | 1292 | 76 | |||
| AK089459.1 | - | 913 | 76 | |||
| AK155135.1 | - | 903 | 76 | |||
| AK171278.1 | - | 6 - 7 | 923 | 76 | ||
| BC051962.1 | - | 6 - 7 | 984 | 76 | ||
| BC086618.1 | - | 6 - 7 | 984 | 76 | ||
| BC132303.1 | - | 5 - 6 | 895 | 40130580 | 76 | |
More Information
Set Membership: |
|

Dfna5
Genomic Map