Assay Details
| Gene Symbol: Dfna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018769.3 | NP_061239.1 | 7 - 8 | 1055 | 105 | |
| GenBank mRNA | AF073309.1 | - | 7 - 8 | 1056 | 105 | |
| AK016561.1 | - | 1423 | 105 | |||
| AK089459.1 | - | 1045 | 105 | |||
| AK155135.1 | - | 1034 | 105 | |||
| AK171278.1 | - | 7 - 8 | 1054 | 105 | ||
| BC132303.1 | - | 6 - 7 | 1026 | 40130580 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dfna5
Genomic Map