| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700000012.mm.1 |
NM_134123.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000012.mm.1 |
NM_134123.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000012.mm.1 |
TC1700000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000010.mm.1 |
NM_134123.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000010.mm.1 |
NM_134123.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000010.mm.1 |
TC1700000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000016.mm.1 |
NM_134123.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000016.mm.1 |
NM_134123.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000016.mm.1 |
TC1700000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000011.mm.1 |
NM_134123.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000011.mm.1 |
NM_134123.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000011.mm.1 |
TC1700000005.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700000015.mm.1 |
NM_134123.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000015.mm.1 |
NM_134123.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700000015.mm.1 |
TC1700000005.mm.1 |
MTA TC |