Assay Details
| Gene Symbol: Scaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134123.3 | NP_598884.2 | 3 - 4 | 570 | 123 | |
| GenBank mRNA | AK122451.1 | - | 3 - 4 | 437 | 123 | |
| AK171078.1 | - | 4 - 5 | 377 | 123 | ||
| AK213588.1 | - | 3 - 4 | 173 | 123 | ||
| AK213688.1 | - | 3 - 4 | 173 | 123 | ||
| AK218304.1 | - | 3 - 4 | 173 | 123 | ||
| BC038363.1 | - | 3 - 4 | 214 | 5371946 | 123 | |
| BC075621.1 | - | 3 - 4 | 563 | 6406441 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scaf8
Genomic Map