Assay Details
| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167860.1 | NP_001161332.1 | 3 - 4 | 389 | 96 | |
| NM_001167861.1 | NP_001161333.1 | 3 - 4 | 389 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wipf3
Genomic Map