Assay Details
| Gene Symbol: Wipf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167860.1 | NP_001161332.1 | 4 - 5 | 1129 | 73 | |
| NM_001167861.1 | NP_001161333.1 | 4 - 5 | 1129 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | NM_001167861.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | AK078889.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | NM_001167860.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | AV348454.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | NM_001167861.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | AK078889.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | NM_001167860.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P316951 | AV348454.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913466 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261552 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80783 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | NM_001167861.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | AK078889.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | NM_001167860.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | AV348454.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | NM_001167861.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | AK078889.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | NM_001167860.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P316951 | AV348454.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01333375 | - | - |
| Whole Mouse Genome G4122A | A_51_P316951 | NM_001167861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316951 | AK078889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316951 | NM_001167860.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316951 | AV348454.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316951 | NM_001167861.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316951 | AK078889.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316951 | NM_001167860.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P316951 | AV348454.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129559 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05007927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05361070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05862468 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05245063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05838770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05245060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463583 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05149031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463619 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05245072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05149037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05245073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05835460 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05007928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05838764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05007926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05838771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463561 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05403985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05007932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05862469 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24463568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10538390 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538392 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10538389 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455301_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10538377 | - | - |
| Mouse Genome 430 2.0 Array | 1455301_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455301_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330319_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 330319_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600004991.mm.1 | NM_001167860.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600004991.mm.1 | NM_001167860.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600004991.mm.1 | NM_001167861.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600004991.mm.1 | NM_001167861.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600004991.mm.1 | TC0600000651.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455301_PM_at | - | - |
| Mouse Expression Set 430 | 1455301_at | - | - |
| Murine Genome U74v2 Set | 165757_PM_i_at | - | - |
| Murine Genome U74v2 Set | 165757_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455301_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455301_at | - | - |

Wipf3
Genomic Map