Assay Details
| Gene Symbol: Slc34a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011392.2 | NP_035522.2 | 1 - 2 | 86 | 91 | |
| GenBank mRNA | CB952922.1 | - | 1 - 2 | 89 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_011392.2 transcripts.
Set Membership: |
|

Slc34a1
Genomic Map