Assay Details
| Gene Symbol: Slc34a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011392.2 | NP_035522.2 | 3 - 4 | 368 | 66 | |
| GenBank mRNA | AB163921.1 | - | 373 | 66 | ||
| AB163922.1 | - | 3 - 4 | 312 | 66 | ||
| AK018740.2 | - | 3 - 4 | 323 | 66 | ||
| BC020529.1 | - | 3 - 4 | 311 | 4223484 | 66 | |
| CB952922.1 | - | 3 - 4 | 371 | 66 | ||
| L33878.1 | - | 2 - 3 | 295 | 66 | ||
| U22465.1 | - | 3 - 4 | 312 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc34a1
Genomic Map