Assay Details
| Gene Symbol: Slc34a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011392.2 | NP_035522.2 | 7 - 8 | 956 | 59 | |
| GenBank mRNA | AB163921.1 | - | 961 | 59 | ||
| AB163922.1 | - | 7 - 8 | 900 | 59 | ||
| AK018740.2 | - | 7 - 8 | 911 | 59 | ||
| BC020529.1 | - | 7 - 8 | 899 | 4223484 | 59 | |
| L33878.1 | - | 6 - 7 | 883 | 59 | ||
| U22465.1 | - | 7 - 8 | 900 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc34a1
Genomic Map