Assay Details
| Gene Symbol: Slc34a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011392.2 | NP_035522.2 | 9 - 10 | 1107 | 54 | |
| GenBank mRNA | AB163921.1 | - | 1112 | 54 | ||
| AB163922.1 | - | 9 - 10 | 1051 | 54 | ||
| AK018740.2 | - | 9 - 10 | 1062 | 54 | ||
| AK032618.1 | - | 2 - 3 | 198 | 54 | ||
| BC020529.1 | - | 9 - 10 | 1050 | 4223484 | 54 | |
| L33878.1 | - | 8 - 9 | 1034 | 54 | ||
| U22465.1 | - | 9 - 10 | 1051 | 54 | ||
More Information
Set Membership: |
|

Slc34a1
Genomic Map