Assay Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293660.1 | NP_001280589.1 | 8 - 9 | 890 | 70 | |
| NM_011307.2 | NP_035437.1 | 10 - 11 | 1728 | 70 | ||
| NR_121592.1 | 9 - 10 | 1647 | 70 | |||
| GenBank mRNA | AK052704.1 | - | 10 - 11 | 1729 | 70 | |
| AK083342.1 | - | 10 - 11 | 1669 | 70 | ||
| AK160926.1 | - | 8 - 9 | 891 | 70 | ||
| BC037092.1 | - | 9 - 10 | 1603 | 70 | ||
| BC040808.1 | - | 10 - 11 | 1684 | 4038778 | 70 | |
| BC051659.1 | - | 10 - 11 | 1651 | 70 | ||
| BC066220.1 | - | 10 - 11 | 1651 | 70 | ||
| U22015.1 | - | 6 - 7 | 1180 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uimc1
Genomic Map