Assay Details
| Gene Symbol: Uimc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293660.1 | NP_001280589.1 | 2 - 3 | 252 | 90 | |
| NM_011307.2 | NP_035437.1 | 2 - 3 | 247 | 90 | ||
| NR_121592.1 | 2 - 3 | 252 | 90 | |||
| GenBank mRNA | AK052704.1 | - | 2 - 3 | 248 | 90 | |
| AK078206.1 | - | 2 - 3 | 251 | 90 | ||
| AK083342.1 | - | 2 - 3 | 188 | 90 | ||
| AK161177.1 | - | 2 - 3 | 251 | 90 | ||
| BC037092.1 | - | 2 - 3 | 208 | 90 | ||
| BC040808.1 | - | 2 - 3 | 203 | 4038778 | 90 | |
| BC051659.1 | - | 2 - 3 | 170 | 90 | ||
| BC066220.1 | - | 2 - 3 | 170 | 90 | ||
More Information
Set Membership: |
|

Uimc1
Genomic Map