Assay Details
| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008498.2 | NP_001008498.2 | 1 - 2 | 197 | 117 | |
| GenBank mRNA | AK014304.1 | - | 1 - 2 | 196 | 117 | |
| BC085113.1 | - | 1 - 2 | 153 | 30730999 | 117 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC085113.1 transcripts.
Set Membership: |
|

Wdr34
Genomic Map