Assay Details
| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008498.2 | NP_001008498.2 | 2 - 3 | 446 | 103 | |
| GenBank mRNA | AK014304.1 | - | 2 - 3 | 445 | 103 | |
| BC085113.1 | - | 2 - 3 | 402 | 30730999 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr34
Genomic Map