Assay Details
| Gene Symbol: Wdr34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008498.2 | NP_001008498.2 | 5 - 6 | 823 | 81 | |
| GenBank mRNA | AK014304.1 | - | 5 - 6 | 822 | 81 | |
| BC085113.1 | - | 5 - 6 | 779 | 30730999 | 81 | |
More Information
Set Membership: |
|

Wdr34
Genomic Map